accessibility menu, dialog, popup

UserName

CAS RN 34941-91-8

IUPAC Name: 2-chloro-4-fluoropyridine
Synonyms: 2-chloro-4-fluoropyridine
Molecular Weight (g/mol): 131.53
Molecular Formula: C5H3ClFN
InChi Key: FGSAQRJRWCZLOB-UHFFFAOYSA-N
SMILES: FC1=CC=NC(Cl)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)