missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoropyridine, 97%

Catalog No. AC442150050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC442150050 5 g Glass bottle
AC442150010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442150050 Supplier Thermo Scientific Chemicals Supplier No. 442150050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34941-91-8 | C5H3ClFN | 131.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34941-91-8
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.54
MDL Number MFCD04039345
InChI Key FGSAQRJRWCZLOB-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-4-fluoro, 2-chloro-4-fluoro pyridine, 2-chloro-4-fluoro-pyridine, pubchem6180, acmc-1aft7, 2-chlro-4-fluoroxypyridine, ksc222e0n, pyrimidine, 4-chloro-6-fluoro, abbypharma ap-14-5731
PubChem CID 2782792
IUPAC Name 2-chloro-4-fluoropyridine
SMILES C1=CN=C(C=C1F)Cl

Specifications

Color Colorless to Yellow
Density 1.4560g/mL
Boiling Point 140°C
Flash Point 54°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.456
Formula Weight 131.54
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-fluoropyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only