accessibility menu, dialog, popup

UserName

OPOOOOPOOOZnZnZn

CAS RN 34807-26-6

IUPAC Name: trizinc(2+) diphosphate
Synonyms: zinc phosphate
Molecular Weight (g/mol): 386.08
Molecular Formula: O8P2Zn3
InChi Key: LRXTYHSAJDENHV-UHFFFAOYSA-H
SMILES: [Zn++].[Zn++].[Zn++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results