missing translation for 'onlineSavingsMsg'
Learn More

Zinc phosphate hydrate, tech.

Catalog No. AA11589A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11589A3 2 kg
AA1158936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA11589A3 Supplier Thermo Scientific Chemicals Supplier No. 011589A3

CAS: 34807-26-6 | O8P2Zn3 | 386.08 g/mol

Zinc phosphate hydrate is used in dentistry. It is commonly used for luting permanent metal restorations and as a base for dental restorations. Zinc phosphate cement is also used for cementation of inlays, crowns, bridges, and orthodontic appliances and occasionally as a temporary restoration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34807-26-6
Molecular Formula O8P2Zn3
Molecular Weight (g/mol) 386.08
MDL Number MFCD00036282
InChI Key LRXTYHSAJDENHV-UHFFFAOYSA-H
Synonym zinc phosphate, delaphos, trizinc diphosphate, zinc orthophosphate, trizinc phosphate, delaphos 2m, bonderite 40, granodine 80, microphos 90, granodine 16nc
PubChem CID 24519

Specifications

Physical Form Powder
Quantity 2 kg
Solubility Information Soluble in acetic acid,dilute mineral acids,ammonia. Insoluble in water
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Zn+2].[Zn+2].[Zn+2]
IUPAC Name trizinc;diphosphate
Formula Weight 386.05 (Anhydrous)
Grade Technical
Chemical Name or Material Zinc phosphate hydrate
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-944-3
RTECSNumber TD0590000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only