accessibility menu, dialog, popup

UserName

H3COCH3OCH3OH

CAS RN 34590-94-8

IUPAC Name: 2-(2-methoxypropoxy)propan-1-ol
Synonyms: 2-(2-methoxypropoxy)propan-1-ol
Molecular Weight (g/mol): 148.2
Molecular Formula: C7H16O3
InChi Key: CUDYYMUUJHLCGZ-UHFFFAOYNA-N
SMILES: COC(C)COC(C)CO
Boiling Point 
  • (2)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)