missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene Glycol Monomethyl Ether (mixture of isomers) 98.0+%, TCI America™

Catalog No. D1053500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1053500G 500 g
D105325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1053500G Supplier TCI America Supplier No. D1053500G

Chemical Identifiers

CAS 34590-94-8
Molecular Formula C7H16O3
MDL Number MFCD00059604

Specifications

Melting Point -80°C
Color Colorless
Boiling Point 188°C
Quantity 500 g
Formula Weight 148.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropylene Glycol Monomethyl Ether (mixture of isomers)
EINECSNumber (2)-0426&(7)-0097
RTECSNumber JM1575000
TSCA Yes