accessibility menu, dialog, popup

UserName

FFFBrFF

CAS RN 344-04-7

IUPAC Name: 1-bromo-2,3,4,5,6-pentafluorobenzene
Synonyms: bromopentafluorobenzene
Molecular Weight (g/mol): 246.96
Molecular Formula: C6BrF5
InChi Key: XEKTVXADUPBFOA-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(Br)C(F)=C1F
Percent Purity 
  • (1)
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (1)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (5)