missing translation for 'onlineSavingsMsg'
Learn More

Bromopentafluorobenzene 99.0+%, TCI America™

Catalog No. B11165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11165G 5 g
B111625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11165G Supplier TCI America Supplier No. B11165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 344-04-7
Molecular Formula C6BrF5
Molecular Weight (g/mol) 246.96
MDL Number MFCD00000287
InChI Key XEKTVXADUPBFOA-UHFFFAOYSA-N
Synonym bromopentafluorobenzene, pentafluorobromobenzene, benzene, bromopentafluoro, bromoperfluorobenzene, pentafluorophenyl bromide, 2,3,4,5,6-pentafluorobromobenzene, benzene, 1-bromo-2,3,4,5,6-pentafluoro, 1-bromo-pentafluorobenzene, 2,3,4,5,6-bromopentafluorobenzene, 1-bromo-2,3,4,5,6-pentafluoro-benzene
PubChem CID 9578
IUPAC Name 1-bromo-2,3,4,5,6-pentafluorobenzene
SMILES FC1=C(F)C(F)=C(Br)C(F)=C1F

Specifications

Melting Point -31°C
Color Yellow
Boiling Point 137°C
Quantity 5 g
Formula Weight 246.96
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromopentafluorobenzene
EINECSNumber (3)-4285
RTECSNumber CY9040552
TSCA Yes