missing translation for 'onlineSavingsMsg'
Learn More

Bromopentafluorobenzene, 99%

Catalog No. AAA1327336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1327336 500 g
AAA1327314 25 g
AAA1327322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1327336 Supplier Thermo Scientific Chemicals Supplier No. A1327336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 344-04-7 | C6BrF5 | 246.96 g/mol

It is used as pharmaceutical intermediate. It is a Grignard reagent (with CuBr in dioxane) can introduce pentafluorophenyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. It is a Grignard reagent (with CuBr in dioxane) can introduce pentafluorophenyl groups.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.

Chemical Identifiers

CAS 344-04-7
Molecular Formula C6BrF5
Molecular Weight (g/mol) 246.96
MDL Number MFCD00000287
InChI Key XEKTVXADUPBFOA-UHFFFAOYSA-N
Synonym bromopentafluorobenzene, pentafluorobromobenzene, benzene, bromopentafluoro, bromoperfluorobenzene, pentafluorophenyl bromide, 2,3,4,5,6-pentafluorobromobenzene, benzene, 1-bromo-2,3,4,5,6-pentafluoro, 1-bromo-pentafluorobenzene, 2,3,4,5,6-bromopentafluorobenzene, 1-bromo-2,3,4,5,6-pentafluoro-benzene
PubChem CID 9578
IUPAC Name 1-bromo-2,3,4,5,6-pentafluorobenzene
SMILES FC1=C(F)C(F)=C(Br)C(F)=C1F

Specifications

Melting Point ∼-31°C
Density 1.981
Boiling Point 136°C to 137°C
Flash Point 87°C (188°F)
Refractive Index 1.45
Quantity 500 g
Beilstein 1819390
Solubility Information Insoluble in water.
Formula Weight 246.97
Percent Purity 99%
Chemical Name or Material Bromopentafluorobenzene
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 206-449-0
RTECSNumber CY9040552
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only