missing translation for 'onlineSavingsMsg'
Learn More

Bromopentafluorobenzene, 99%

Catalog No. AC107221000
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107221000 100 mL Glass bottle
AC107220050 5 mL Glass bottle
AC107220250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107221000 Supplier Thermo Scientific Chemicals Supplier No. 107221000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 344-04-7 | C6BrF5 | 246.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 344-04-7
Molecular Formula C6BrF5
Molecular Weight (g/mol) 246.96
MDL Number MFCD00000287
InChI Key XEKTVXADUPBFOA-UHFFFAOYSA-N
Synonym bromopentafluorobenzene, pentafluorobromobenzene, benzene, bromopentafluoro, bromoperfluorobenzene, pentafluorophenyl bromide, 2,3,4,5,6-pentafluorobromobenzene, benzene, 1-bromo-2,3,4,5,6-pentafluoro, 1-bromo-pentafluorobenzene, 2,3,4,5,6-bromopentafluorobenzene, 1-bromo-2,3,4,5,6-pentafluoro-benzene
PubChem CID 9578
IUPAC Name 1-bromo-2,3,4,5,6-pentafluorobenzene
SMILES FC1=C(F)C(F)=C(Br)C(F)=C1F

Specifications

Melting Point -31°C
Color Colorless
Density 1.9810g/mL
Boiling Point 137°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4485 to 1.4505
Linear Formula BrC6F5
Quantity 100 mL
Specific Gravity 1.981
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diethyl ether,benzene
Formula Weight 246.96
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Bromopentafluorobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-449-

RUO – Research Use Only