accessibility menu, dialog, popup

UserName

CAS RN 3430-14-6

CH3NH2N

CAS RN 3430-14-6

IUPAC Name: 6-methylpyridin-3-amine
Synonyms: 6-methylpyridin-3-amine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: UENBBJXGCWILBM-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)