missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methylpyridine 98.0+%, TCI America™

Catalog No. A20535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20535G 5 g
A20531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20535G Supplier TCI America Supplier No. A20535G
Only null left

Chemical Identifiers

CAS 3430-14-6
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00833389
InChI Key UENBBJXGCWILBM-UHFFFAOYSA-N
Synonym 5-amino-2-methylpyridine, 3-amino-6-methylpyridine, 3-amino-6-picoline, 5-amino-2-picoline, 6-methyl-pyridin-3-ylamine, 2-methyl-5-aminopyridine, 6-methylpyridin-3-ylamine, 3-pyridinamine, 6-methyl, 6-methyl-3-pyridinamine, 6-methyl-3-pyridylamine
PubChem CID 243313
IUPAC Name 6-methylpyridin-3-amine
SMILES CC1=CC=C(N)C=N1

Specifications

Melting Point 95°C
Quantity 5 g
UN Number 2811
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-methylpyridine