accessibility menu, dialog, popup

UserName

H3CONH2O

CAS RN 3424-93-9

IUPAC Name: 4-methoxybenzamide
Synonyms: P-methyoxybenzamide
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: GUCPYIYFQVTFSI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(N)=O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)