accessibility menu, dialog, popup

UserName

CAS RN 34006-16-1

HClSHN

CAS RN 34006-16-1

IUPAC Name: hydrogen quinoline-8-thiol chloride
Synonyms: hydrogen 8-mercaptoquinoline chloride
Molecular Weight (g/mol): 197.68
Molecular Formula: C9H8ClNS
InChi Key: RWBSBQAUAJSGHY-UHFFFAOYSA-N
SMILES: [H+].[Cl-].SC1=C2N=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results