missing translation for 'onlineSavingsMsg'
Learn More

8-Mercaptoquinoline Hydrochloride 95.0+%, TCI America™

Catalog No. A50045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50045G 5 g
A50041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50045G Supplier TCI America Supplier No. A50045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34006-16-1
Molecular Formula C9H8ClNS
Molecular Weight (g/mol) 197.68
MDL Number MFCD00043261
InChI Key RWBSBQAUAJSGHY-UHFFFAOYSA-N
Synonym Thiooxine Hydrochloride
PubChem CID 3082386
IUPAC Name hydrogen quinoline-8-thiol chloride
SMILES [H+].[Cl-].SC1=C2N=CC=CC2=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 197.68
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Mercaptoquinoline Hydrochloride [Extraction-spectrophotometric and fluorimetric reagent for soft m