accessibility menu, dialog, popup

UserName

HOO

CAS RN 3400-45-1

IUPAC Name: cyclopentanecarboxylic acid
Synonyms: cyclopentanecarboxylic acid
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: JBDSSBMEKXHSJF-UHFFFAOYSA-N
SMILES: OC(=O)C1CCCC1
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)