missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarboxylic acid, 99%

Catalog No. AAA1237530
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1237530 250 g
AAA1237509 10 g
AAA1237518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1237530 Supplier Thermo Scientific Chemicals Supplier No. A1237530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3400-45-1 | C6H10O2 | 114.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3400-45-1
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00001371
InChI Key JBDSSBMEKXHSJF-UHFFFAOYSA-N
Synonym cyclopentane carboxylic acid, cyclopentylcarboxylic acid, cyclopentancarboxylic acid, cyclopentanoic acid, unii-z6691vh94a, cyclopentylmethanoic acid, pubchem14153, carboxycyclopentane, cyclopentyl carboxylic acid, acmc-209i4g
PubChem CID 18840
IUPAC Name cyclopentanecarboxylic acid
SMILES C1CCC(C1)C(=O)O

Specifications

Melting Point -7°C
Density 1.052
Boiling Point 215°C to 216°C
Flash Point 93°C (199°F)
Refractive Index 1.454
Quantity 250 g
Beilstein 2039729
Formula Weight 114.14
Percent Purity 99%
Chemical Name or Material Cyclopentanecarboxylic acid
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 222-269-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only