accessibility menu, dialog, popup

UserName

CAS RN 340-07-8

FFFON

CAS RN 340-07-8

IUPAC Name: 2,2,2-trifluoro-1-(piperidin-1-yl)ethan-1-one
Synonyms: 2,2,2-trifluoro-1-(piperidin-1-yl)ethanone
Molecular Weight (g/mol): 181.16
Molecular Formula: C7H10F3NO
InChi Key: BCJUMGSHTLYECD-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)N1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results