CAS RN 33898-90-7
CAS RN 33898-90-7
IUPAC Name:
3-oxo-3-(thiophen-2-yl)propanenitrile
Synonyms:
3-oxo-3-(thiophen-2-yl)propanenitrile
Molecular Weight (g/mol):
151.18
Molecular Formula:
C7H5NOS
InChi Key:
XWWUQBHVRILEPB-UHFFFAOYSA-N
SMILES:
O=C(CC#N)C1=CC=CS1
1
–
1
of
1
results
1
3-Oxo-3-(2-thienyl)propionitrile, 98%
CAS: 33898-90-7 Molecular Formula: C7H5NOS Molecular Weight (g/mol): 151.18 MDL Number: MFCD00052586 InChI Key: XWWUQBHVRILEPB-UHFFFAOYSA-N Synonym: 2-thenoylacetonitrile,3-oxo-3-thiophen-2-yl propanenitrile,3-oxo-3-2-thienyl propanenitrile,3-oxo-3-2-thienyl propionitrile,2-thiophene-2-carbonyl acetonitrile,3-oxo-3-thiophen-2-yl-propionitrile,3-2-thienyl-3-oxopropanenitrile,pubchem8372,then-2-oylacetonitrile,acmc-1csfg PubChem CID: 141853 IUPAC Name: 3-oxo-3-thiophen-2-ylpropanenitrile SMILES: O=C(CC#N)C1=CC=CS1