missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-3-(2-thienyl)propionitrile, 98%

Catalog No. AAA1091114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1091114 25 g
AAA1091103 1 g
AAA1091106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1091114 Supplier Thermo Scientific Chemicals Supplier No. A1091114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33898-90-7 | C7H5NOS | 151.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33898-90-7
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.18
MDL Number MFCD00052586
InChI Key XWWUQBHVRILEPB-UHFFFAOYSA-N
Synonym 2-thenoylacetonitrile, 3-oxo-3-thiophen-2-yl propanenitrile, 3-oxo-3-2-thienyl propanenitrile, 3-oxo-3-2-thienyl propionitrile, 2-thiophene-2-carbonyl acetonitrile, 3-oxo-3-thiophen-2-yl-propionitrile, 3-2-thienyl-3-oxopropanenitrile, pubchem8372, then-2-oylacetonitrile, acmc-1csfg
PubChem CID 141853
IUPAC Name 3-oxo-3-thiophen-2-ylpropanenitrile
SMILES O=C(CC#N)C1=CC=CS1

Specifications

Melting Point 132°C to 136°C
Quantity 25 g
UN Number UN3439
Beilstein 111990
Formula Weight 151.19
Percent Purity 98%
Chemical Name or Material 3-Oxo-3- (2-thienyl)propionitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only