accessibility menu, dialog, popup

UserName

CAS RN 335421-90-4

PSOOSOOSOOOOONaNaNa

CAS RN 335421-90-4

IUPAC Name: trisodium 3-[bis(3-sulfonatophenyl)phosphanyl]benzene-1-sulfonate
Synonyms: tppts
Molecular Weight (g/mol): 568.41
Molecular Formula: C18H12Na3O9PS3
InChi Key: MYAJTCUQMQREFZ-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC(=CC=C1)P(C1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=CC(=C1)S([O-])(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results