accessibility menu, dialog, popup

UserName

FFFFFFFFFFFFFFFClO

CAS RN 335-64-8

IUPAC Name: pentadecafluorooctanoyl chloride
Synonyms: pentadecafluorooctanoyl chloride
Molecular Weight (g/mol): 432.51
Molecular Formula: C8ClF15O
InChi Key: AQQBRCXWZZAFOK-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)