missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctanoyl chloride, 97%, Thermo Scientific™

Catalog No. AAL0929414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0929414 25 g
AAL0929406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0929414 Supplier Thermo Scientific Chemicals Supplier No. L0929414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Perfluorooctanoyl chloride is derivatizated and used in the determination of Amphetamine and Methamphetamine in Blood. It can be used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from oxidizing agents and moisture. Store under dry inert gas. Incompatible with strong oxidizing agents, strong bases, alcohols.

Chemical Identifiers

CAS 335-64-8
Molecular Formula C8ClF15O
Molecular Weight (g/mol) 432.51
MDL Number MFCD00013654
InChI Key AQQBRCXWZZAFOK-UHFFFAOYSA-N
Synonym perfluorooctanoyl chloride, pentadecafluorooctanoyl chloride, pentadecafluorooctyl chloride, octanoyl chloride, pentadecafluoro, perfluorooctanoylchloride, perfluorooctanyl chloride, perflurooctanoyl chloride, acmc-1cmee, perfluorocaprylic chloride, n-perfluorooctanoyl chloride
PubChem CID 78978
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(Cl)=O

Specifications

Density 1.744
Boiling Point 130°C to 132°C
Refractive Index 1.3045
Quantity 25 g
UN Number UN3265
Beilstein 1809677
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
IUPAC Name pentadecafluorooctanoyl chloride
Formula Weight 432.52
Percent Purity 97%
Chemical Name or Material Perfluorooctanoyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-394-2
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen