accessibility menu, dialog, popup

UserName

CAS RN 334658-75-2

IUPAC Name: (10-phenylanthracen-9-yl)boronic acid
Synonyms: 10-phenylanthracen-9-ylboronic acid
Molecular Weight (g/mol): 298.15
Molecular Formula: C20H15BO2
InChi Key: RVPCPPWNSMAZKR-UHFFFAOYSA-N
SMILES: OB(O)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12
Physical Form 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)