missing translation for 'onlineSavingsMsg'
Learn More

10-Phenyl-9-anthraceneboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. P19841G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19841G Supplier TCI America Supplier No. P19841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 334658-75-2
Molecular Formula C20H15BO2
Molecular Weight (g/mol) 298.15
MDL Number MFCD11111989
InChI Key RVPCPPWNSMAZKR-UHFFFAOYSA-N
Synonym 10-phenylanthracen-9-yl boronic acid, 10-phenyl-9-anthracene boronic acid, 10-phenyl-9-anthraceneboronic acid, 10-phenylanthracene-9-boronic acid, 10-phenylantrhacen-9-yl boronic acid, boronic acid, 10-phenyl-9-anthracenyl, pubchem19639, 9-borono-10-phenylanthracene, 10-phenylanthracene-9-ylboronic acid, 10-phenyl-9-anthryl boronic acid
PubChem CID 22247164
IUPAC Name (10-phenylanthracen-9-yl)boronic acid
SMILES OB(O)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12

Specifications

Color Yellow
Quantity 1 g
Formula Weight 298.15
Physical Form Crystalline Powder
Chemical Name or Material 10-Phenyl-9-anthraceneboronic Acid (contains varying amounts of Anhydride)