accessibility menu, dialog, popup

UserName

LiOHSOOFFF

CAS RN 33454-82-9

IUPAC Name: trifluoromethanesulfonic acid lithium
Synonyms: trifluoromethanesulfonic acid, lithium salt
Molecular Weight (g/mol): 157.01
Molecular Formula: CHF3LiO3S
InChi Key: WDGKXRCNMKPDSD-UHFFFAOYSA-N
SMILES: [Li].OS(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)