missing translation for 'onlineSavingsMsg'
Learn More

Lithium trifluoromethanesulfonate, 97%

Catalog No. AA3932218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3932218 50 g
AA3932209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3932218 Supplier Thermo Scientific Chemicals Supplier No. 03932218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 33454-82-9 | CF3LiO3S | 156.003 g/mol

Lithium trifluoromethanesulfonate is mixed with ceramic filler to prepare polyethylene oxide films in order to improve the electrochemical and mechanical characteristics of the membrane, which is used in the lithium batteries. It acts as a doping salt used in the preparation of nano manganese-composite polymer electrolytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33454-82-9
Molecular Formula CF3LiO3S
Molecular Weight (g/mol) 156.003
MDL Number MFCD00013227
InChI Key MCVFFRWZNYZUIJ-UHFFFAOYSA-M
Synonym lithium trifluoromethanesulfonate, lithium triflate, li triflate, lithium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, lithium salt, unii-1c966kv50i, trifluoromethanesulfonic acid lithium salt, lithium 1+ ion trifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, lithium salt 1:1, trifluoromethanesulfonic acid, lithium salt
PubChem CID 3664839
IUPAC Name lithium;trifluoromethanesulfonate
SMILES [Li+].C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 423°C
Color White
Quantity 50 g
Sensitivity Moisture sensitive
Solubility Information Miscible with water.
Formula Weight 156.01
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Lithium trifluoromethanesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 251-528-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only