CAS RN 3344-18-1
IUPAC Name:
bis(2-hydroxypropane-1,2,3-tricarboxylic acid) trimagnesium
Synonyms:
magnesium citrate (3:2)
Molecular Weight (g/mol):
457.16
Molecular Formula:
C12H16Mg3O14
InChi Key:
AOHMFUYIHARAGR-UHFFFAOYSA-N
SMILES:
[Mg].[Mg].[Mg].OC(=O)CC(O)(CC(O)=O)C(O)=O.OC(=O)CC(O)(CC(O)=O)C(O)=O
1
–
2
of
2
results
1
Magnesium citrate hydrate, Mg ca 16% (dry wt.)
CAS: 3344-18-1 Molecular Formula: C12H12Mg3O15 Molecular Weight (g/mol): 469.128 MDL Number: MFCD09953657 InChI Key: HZTVQLXIRHEMMB-UHFFFAOYSA-H Synonym: trimagnesium 2+ dicitrate hydrate PubChem CID: 122198783 IUPAC Name: trimagnesium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[Mg+2].[Mg+2].[Mg+2]
2
Magnesium Citrate, Anhydrous, USP, 14.5-16.4%, Spectrum™ Chemical
CAS: 3344-18-1 Molecular Formula: C12H16Mg3O14 Molecular Weight (g/mol): 457.16 InChI Key: AOHMFUYIHARAGR-UHFFFAOYSA-N IUPAC Name: bis(2-hydroxypropane-1,2,3-tricarboxylic acid) trimagnesium SMILES: [Mg].[Mg].[Mg].OC(=O)CC(O)(CC(O)=O)C(O)=O.OC(=O)CC(O)(CC(O)=O)C(O)=O