missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Citrate, Anhydrous, USP, 14.5-16.4%, Spectrum™ Chemical

Catalog No. 18604057
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604057 50 kg Fiber Drum
18604053 125 g Poly Bottle
18604054 500 g Poly Bottle
18604055 2.5 kg Poly Pail
18604056 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604057 Supplier Spectrum Chemical Mfg Cor Supplier No. M203650KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M2036, 3344-18-1, C12H10Mg3O14

Spectrum™ Chemical Magnesium Citrate, Anhydrous, USP is often used as a saline laxative to clean stool from the intestines before surgery or certain bowel procedures. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 3344-18-1
Molecular Formula C12H16Mg3O14
Molecular Weight (g/mol) 457.16
InChI Key AOHMFUYIHARAGR-UHFFFAOYSA-N
IUPAC Name bis(2-hydroxypropane-1,2,3-tricarboxylic acid) trimagnesium
SMILES [Mg].[Mg].[Mg].OC(=O)CC(O)(CC(O)=O)C(O)=O.OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

pH 5.0-9.0
CAS Max % 100%
Quantity 50 kg
Percent Purity 14.5 to 16.4%
Grade USP
Loss on Drying 2%
Packaging Fiber Drum