accessibility menu, dialog, popup

UserName

CAS RN 33414-62-9

IUPAC Name: 2-(oxolan-2-yl)acetonitrile
Synonyms: 2-(oxolan-2-yl)acetonitrile
Molecular Weight (g/mol): 111.14
Molecular Formula: C6H9NO
InChi Key: LSCVJPYMXOAEPX-UHFFFAOYNA-N
SMILES: N#CCC1CCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results