missing translation for 'onlineSavingsMsg'
Learn More

(Tetrahydrofuran-2-yl)acetonitrile 98.0+%, TCI America™

Catalog No. T26225G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T26225G Supplier TCI America Supplier No. T26225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33414-62-9
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.144
MDL Number MFCD03206782
InChI Key LSCVJPYMXOAEPX-UHFFFAOYSA-N
Synonym Tetrahydrofurfurylcarbonitrile, 2-(Cyanomethyl)tetrahydrofuran
PubChem CID 4962721
IUPAC Name 2-(oxolan-2-yl)acetonitrile
SMILES C1CC(OC1)CC#N

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
Formula Weight 111.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Tetrahydrofuran-2-yl)acetonitrile