accessibility menu, dialog, popup

UserName

CAS RN 3339-73-9

IUPAC Name: 3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid
Synonyms: 3-(1,3-dioxoisoindol-2-yl)propanoic acid
Molecular Weight (g/mol): 219.2
Molecular Formula: C11H9NO4
InChi Key: DXXHRZUOTPMGEH-UHFFFAOYSA-N
SMILES: OC(=O)CCN1C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)