missing translation for 'onlineSavingsMsg'
Learn More

3-Phthalimidopropionic acid, 98%

Catalog No. AAL1353506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1353506 5 g
AAL1353503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1353506 Supplier Thermo Scientific Chemicals Supplier No. L1353506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 3339-73-9 | C11H9NO4 | 219.196 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3339-73-9
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.196
MDL Number MFCD00023096
InChI Key DXXHRZUOTPMGEH-UHFFFAOYSA-N
Synonym 3-phthalimidopropionic acid, 3-1,3-dioxoisoindolin-2-yl propanoic acid, 2h-isoindole-2-propanoic acid, 1,3-dihydro-1,3-dioxo, 3-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl propanoic acid, 3-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl propanoic acid, pht-beta-ala-oh, b-phthalimidopropionic acid, 3-phthalimidopropanoic acid, n-phthaloyl-beta-alanine, phthalyl-.beta.-alanine
PubChem CID 76859
IUPAC Name 3-(1,3-dioxoisoindol-2-yl)propanoic acid
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCC(=O)O

Specifications

Melting Point 151°C to 153°C
Quantity 5 g
Beilstein 15570
Formula Weight 219.2
Percent Purity 98%
Chemical Name or Material 3-Phthalimidopropionic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 222-083-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only