accessibility menu, dialog, popup

UserName

ClCH3O(S)SNNH3CCH3O(S)OH3COH

CAS RN 33286-22-5

IUPAC Name: hydrogen (2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl acetate chloride
Synonyms: hydrogen diltiazem chloride
Molecular Weight (g/mol): 450.98
Molecular Formula: C22H27ClN2O4S
InChi Key: HDRXZJPWHTXQRI-BHDTVMLSSA-N
SMILES: [H+].[Cl-].COC1=CC=C(C=C1)[C@@H]1SC2=CC=CC=C2N(CCN(C)C)C(=O)[C@@H]1OC(C)=O
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Physical Form 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (5)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (1)