missing translation for 'onlineSavingsMsg'
Learn More

(+)-cis-Diltiazem hydrochloride

Catalog No. AAJ6324514
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6324514 25 g
AAJ6324506 5 g
AAJ6324509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6324514 Supplier Thermo Scientific Chemicals Supplier No. J6324514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An L-type calcium channel blocker | CAS: 33286-22-5 | C22H27ClN2O4S | 450.978 g/mol

An L-type calcium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33286-22-5
Molecular Formula C22H27ClN2O4S
Molecular Weight (g/mol) 450.978
MDL Number MFCD00069252
InChI Key HDRXZJPWHTXQRI-BHDTVMLSSA-N
Synonym diltiazem hydrochloride, diltiazem hcl, herbesser, dilzene, lacerol, masdil, tildiem, tiazac, mono-tildiem, bi-tildiem
PubChem CID 62920
ChEBI CHEBI:645509
IUPAC Name [(2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochloride
SMILES CC(=O)OC1C(SC2=CC=CC=C2N(C1=O)CCN(C)C)C3=CC=C(C=C3)OC.Cl

Specifications

Melting Point 207°C to 212°C
Color White
Quantity 25 g
Beilstein 4228706
Merck Index 14,3202
Solubility Information Soluble in water at 50mg/ml
Formula Weight 450.98
Physical Form Powder
Chemical Name or Material (+)-cis-Diltiazem hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 251-443-3
RTECSNumber DL0310000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only