accessibility menu, dialog, popup

UserName

CAS RN 3328-70-9

OHOO

CAS RN 3328-70-9

IUPAC Name: 4-hydroxybenzene-1,3-dicarbaldehyde
Synonyms: 4-hydroxybenzene-1,3-dicarbaldehyde
Molecular Weight (g/mol): 150.13
Molecular Formula: C8H6O3
InChi Key: FEUATHOQKVGPEK-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=O)C=C1C=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results