accessibility menu, dialog, popup

UserName

CAS RN 331771-20-1

CH3ONNHNHNOONO

CAS RN 331771-20-1

IUPAC Name: N-[4-({6-methoxy-7-[3-(morpholin-4-yl)propoxy]quinazolin-4-yl}amino)phenyl]benzamide
Synonyms: N-[4-({6-methoxy-7-[3-(morpholin-4-yl)propoxy]quinazolin-4-yl}amino)phenyl]benzamide
Molecular Weight (g/mol): 513.6
Molecular Formula: C29H31N5O4
InChi Key: OGNYUTNQZVRGMN-UHFFFAOYSA-N
SMILES: COC1=C(OCCCN2CCOCC2)C=C2N=CN=C(NC3=CC=C(NC(=O)C4=CC=CC=C4)C=C3)C2=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results