missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Aurora Kinase Inhibitor VI, ZM447439
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | White |
| Description | 98% by HPLC |
| Product Type | Inhibitor |
| CAS | 331771-20-1 |
| Molecular Formula | C29H31N5O4 |
| Formula Weight | 513.6g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 5 mg |