CAS RN 33005-95-7
IUPAC Name:
2-(5-benzoylthiophen-2-yl)propanoic acid
Synonyms:
tiaprofenic acid nu-tiaprofenic apo-tiaprofenic novo-tiaprofenic tiaprofenic-300 - tab flanid tiaprofenic-200 - tab tiaprofensaeure apo-tiaprofenic tablets florizel brand of tiaprofenic acid
Molecular Weight (g/mol):
260.31
Molecular Formula:
C14H12O3S
InChi Key:
GUHPRPJDBZHYCJ-UHFFFAOYNA-N
SMILES:
CC(C(O)=O)C1=CC=C(S1)C(=O)C1=CC=CC=C1
1
–
2
of
2
results
1
Thermo Scientific Chemicals Tiaprofenic acid
CAS: 33005-95-7 Molecular Formula: C14H12O3S
2
Tiaprofenic Acid 98.0+%, TCI America™
CAS: 33005-95-7 Molecular Formula: C14H12O3S Molecular Weight (g/mol): 260.31 MDL Number: MFCD00866089 InChI Key: GUHPRPJDBZHYCJ-UHFFFAOYNA-N Synonym: 2-(5-Benzoylthiophen-2-yl)propionic Acid PubChem CID: 5468 ChEBI: CHEBI:32221 IUPAC Name: 2-(5-benzoylthiophen-2-yl)propanoic acid SMILES: CC(C(O)=O)C1=CC=C(S1)C(=O)C1=CC=CC=C1