CAS RN 330-17-6
IUPAC Name:
4-[(trifluoromethyl)sulfanyl]benzoate
Synonyms:
4-[(trifluoromethyl)sulfanyl]benzoate
Molecular Weight (g/mol):
221.17
Molecular Formula:
C8H4F3O2S
InChi Key:
UMOGQQWVQUQTQA-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=C(SC(F)(F)F)C=C1
1
–
1
of
1
results
1
4-(Trifluoromethylthio)benzoic Acid 97.0+%, TCI America™
CAS: 330-17-6 Molecular Formula: C8H5F3O2S Molecular Weight (g/mol): 222.181 MDL Number: MFCD00040906 InChI Key: UMOGQQWVQUQTQA-UHFFFAOYSA-N Synonym: 4-trifluoromethylthio benzoic acid,4-trifluoromethyl sulfanyl benzoic acid,4-trifluoromethyl thio benzoic acid,4-trifluoromethylsulfanyl-benzoic acid,4-trifluoromethylsulfanyl benzoic acid,4-trifluoromethyl-mercapto-benzoic acid,pubchem13674,acmc-209hxv,4-trifluoromethylthiobenzoic acid PubChem CID: 2777858 IUPAC Name: 4-(trifluoromethylsulfanyl)benzoic acid SMILES: C1=CC(=CC=C1C(=O)O)SC(F)(F)F