missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)benzoic Acid 97.0+%, TCI America™

Catalog No. T23905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23905G 5 g
T239025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23905G Supplier TCI America Supplier No. T23905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 330-17-6
Molecular Formula C8H5F3O2S
Molecular Weight (g/mol) 222.181
MDL Number MFCD00040906
InChI Key UMOGQQWVQUQTQA-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio benzoic acid, 4-trifluoromethyl sulfanyl benzoic acid, 4-trifluoromethyl thio benzoic acid, 4-trifluoromethylsulfanyl-benzoic acid, 4-trifluoromethylsulfanyl benzoic acid, 4-trifluoromethyl-mercapto-benzoic acid, pubchem13674, acmc-209hxv, 4-trifluoromethylthiobenzoic acid
PubChem CID 2777858
IUPAC Name 4-(trifluoromethylsulfanyl)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)SC(F)(F)F

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 5 g
Formula Weight 222.18
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethylthio)benzoic Acid