accessibility menu, dialog, popup

UserName

NHOPOOHHOPOHOONHOPOOHHOPOHOOH2OH2OH2OH2OH2ONaNa

CAS RN 329003-65-8

IUPAC Name: disodium pentahydrate bis(hydrogen [1-hydroxy-1-phosphono-2-(pyridin-3-yl)ethyl]phosphonate)
Synonyms: disodium pentahydrate bis(hydrogen 1-hydroxy-1-phosphono-2-(pyridin-3-yl)ethylphosphonate)
Molecular Weight (g/mol): 700.26
Molecular Formula: C14H30N2Na2O19P4
InChi Key: HYFDYHPNTXOPPO-UHFFFAOYNA-L
SMILES: O.O.O.O.O.[Na+].[Na+].OC(CC1=CN=CC=C1)(P(O)(O)=O)P(O)([O-])=O.OC(CC1=CN=CC=C1)(P(O)(O)=O)P(O)([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results