missing translation for 'onlineSavingsMsg'
Learn More

Risedronate Sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013937
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013937 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013937 Supplier Merck Emd Millipore Supplier No. PHR18881G

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 329003-65-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C7H10 NNaO7P2 · 2.5H2 O
MDL Number MFCD01706268
Synonym Risedronic acid monosodium salt hemi(pentahydrate)
Type Certified Reference Material
Quantity 1 g
Formula Weight 350.13
Grade Certified Reference Material