accessibility menu, dialog, popup

UserName

CAS RN 32634-68-7

IUPAC Name: (2S,3S)-2,3-bis(4-methylbenzoyloxy)butanedioic acid
Synonyms: (+)-ditoluoyltartaric acid
Molecular Weight (g/mol): 386.36
Molecular Formula: C20H18O8
InChi Key: CMIBUZBMZCBCAT-HOTGVXAUSA-N
SMILES: CC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results