missing translation for 'onlineSavingsMsg'
Learn More

(+)-Di-p-toluoyl-D-tartaric Acid 98.0+%, TCI America™

Catalog No. D141725G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D141725G 25 g
D1417250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D141725G Supplier TCI America Supplier No. D141725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32634-68-7 | C20H18O8 | 386.36 g/mol

Chemical Identifiers

CAS 32634-68-7
Molecular Formula C20H18O8
Molecular Weight (g/mol) 386.36
MDL Number MFCD00008552
InChI Key CMIBUZBMZCBCAT-HOTGVXAUSA-N
Synonym di-p-toluoyl-d-tartaric acid, +-di-1,4-toluoyl-d-tartaric acid, 2s,3s-2,3-bis 4-methylbenzoyl oxy succinic acid, unii-4480ma5qip, ditoluoyltartaric acid, +, +-dtta, butanedioic acid, 2,3-bis 4-methylbenzoyl oxy-, 2s,3s, 2s,3s-2,3-bis 4-methylbenzoyl oxy butanedioic acid, --di-p-toluoyl-l-trataric acid
PubChem CID 263211
IUPAC Name (2S,3S)-2,3-bis(4-methylbenzoyloxy)butanedioic acid
SMILES CC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O

Specifications

Melting Point 171°C
Color White
Quantity 25 g
Formula Weight 386.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Di-p-toluoyl-D-tartaric Acid