missing translation for 'onlineSavingsMsg'
Learn More

Rivastigmine Related Compound A, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111013931
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013931 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013931 Supplier Merck Emd Millipore Supplier No. PHR202930MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 32634-68-7
Linear Formula [CH3C6H4CO2CH(CO2H)-]2
MDL Number MFCD00008552
Melting Point 169°C to 171°C
Synonym (+)-O,O'-Di-p-toluoyl-D-tartaric acid; Di-p-toluoyl-D-tartaric acid
Quantity 30 mg
Formula Weight 386.35
Grade Certified Reference Material