accessibility menu, dialog, popup

UserName

HNNNH2OHSOOHO(E/Z)

CAS RN 32514-47-9

IUPAC Name: N,N''-diphenylguanidine; sulfuric acid
Synonyms: 1,3-diphenylguanidine; sulfuric acid
Molecular Weight (g/mol): 309.34
Molecular Formula: C13H15N3O4S
InChi Key: UYSNRRIIIXZGOZ-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC(NC1=CC=CC=C1)=NC1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results