missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylguanidine Sulfate, TCI America™

Catalog No. D134825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D134825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D134825G Supplier TCI America Supplier No. D134825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32514-47-9
Molecular Formula C13H15N3O4S
Molecular Weight (g/mol) 309.34
MDL Number MFCD00060241
InChI Key UYSNRRIIIXZGOZ-UHFFFAOYSA-N
PubChem CID 57377215
IUPAC Name N,N''-diphenylguanidine; sulfuric acid
SMILES OS(O)(=O)=O.NC(NC1=CC=CC=C1)=NC1=CC=CC=C1

Specifications

Color White
Quantity 25 g
Formula Weight 520.61
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diphenylguanidine Sulfate