CAS RN 32477-35-3
IUPAC Name:
2,2,3,3,4,4,4-heptafluoro-1-(1H-imidazol-1-yl)butan-1-one
Synonyms:
2,2,3,3,4,4,4-heptafluoro-1-(imidazol-1-yl)butan-1-one
Molecular Weight (g/mol):
264.1
Molecular Formula:
C7H3F7N2O
InChi Key:
MSYHGYDAVLDKCE-UHFFFAOYSA-N
SMILES:
FC(F)(F)C(F)(F)C(F)(F)C(=O)N1C=CN=C1
1
–
1
of
1
results
1
1-(Heptafluorobutyryl)imidazole 97.0+%, TCI America™
CAS: 32477-35-3 Molecular Formula: C7H3F7N2O Molecular Weight (g/mol): 264.103 MDL Number: MFCD00014503 InChI Key: MSYHGYDAVLDKCE-UHFFFAOYSA-N Synonym: n-heptafluorobutyrylimidazole,1-heptafluorobutyryl imidazole,heptafluorobutyrylimidazole,n-heptafluorobutyryl imidazole,hfbi,1-perfluorobutyryl imidazole,2,2,3,3,4,4,4-heptafluoro-1-1h-imidazol-1-yl butan-1-one,1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl-1h-imidazole,2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-yl butan-1-one,1h-imidazole, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl PubChem CID: 94431 IUPAC Name: 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-ylbutan-1-one SMILES: C1=CN(C=N1)C(=O)C(C(C(F)(F)F)(F)F)(F)F