missing translation for 'onlineSavingsMsg'
Learn More

1-(Heptafluorobutyryl)imidazole 97.0+%, TCI America™

Catalog No. H04675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04675G 5 g
H046725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04675G Supplier TCI America Supplier No. H04675G

Chemical Identifiers

CAS 32477-35-3
Molecular Formula C7H3F7N2O
Molecular Weight (g/mol) 264.103
MDL Number MFCD00014503
InChI Key MSYHGYDAVLDKCE-UHFFFAOYSA-N
Synonym n-heptafluorobutyrylimidazole, 1-heptafluorobutyryl imidazole, heptafluorobutyrylimidazole, n-heptafluorobutyryl imidazole, hfbi, 1-perfluorobutyryl imidazole, 2,2,3,3,4,4,4-heptafluoro-1-1h-imidazol-1-yl butan-1-one, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl-1h-imidazole, 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-yl butan-1-one, 1h-imidazole, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl
PubChem CID 94431
IUPAC Name 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-ylbutan-1-one
SMILES C1=CN(C=N1)C(=O)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Melting Point 13°C
Color Yellow
Boiling Point 161°C
Quantity 5 g
Formula Weight 264.10
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(Heptafluorobutyryl)imidazole [Acylating Agent]
TSCA No
Recommended Storage Refrigerator